Reagents Sulfuric Acid, ACS Reagent, Winpak Container, 75 lb

Representative image only.

Reagents Sulfuric Acid, ACS Reagent, Winpak Container, 75 lb

Reagents | Mfr: C5102600-75LB| Item: 6228647

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Assay 95 to 98%
  • Colorless liquid
  • Density: 1.84
  • Solubility information: miscible
  • pH range: <1
  • 75 lb Winpak container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaH2SO4
  • ClassAcid
  • FormLiquid
  • Weight (mL)34019
  • Formula Weight98.0785
  • PackagingWinpak container
  • DOT InformationSULFURIC ACID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number7664-93-9
  • UN NumberUN1830
  • Shelf Life (days)1125
  • Shelf Life (months)60
  • Boiling Point C (° C)335
  • Boiling Point F (° F)635
  • Melting Point C (° C)-20
  • Melting Point F (° F)-4
  • DescriptionSulfuric Acid, ACS Reagent, Winpak Container, 75 lb
Documents & Certificates
Reviews
Q&A