Reagents® Sulfuric Acid 70% (w/w), Textile Grade

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless to tan liquid
  • Density: 1.61
  • Solubility information: miscible
  • pH range: <0.1
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Description
Availability Pricing
Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Drum, 15 gal. 6228825 Mfr # C7306600-15E
COMPARE
Volumetric Drum Sulfuric Acid, 70% (w/w), Textile Grade, Drum, 15 gal.
Call For Price
Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Natural Poly Bottle, 1 L 6228826 Mfr # C7306600-1A
COMPARE
Volumetric Poly bottle Sulfuric Acid, 70% (w/w), Textile Grade, Natural Poly Bottle, 1 L
Call For Price
Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Drum, 55 gal. 6228827 Mfr # C7306600-55E
COMPARE
Volumetric Drum Sulfuric Acid, 70% (w/w), Textile Grade, Drum, 55 gal.
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat