Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Drum, 15 gal.

Representative image only.

Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Drum, 15 gal.

Reagents | Mfr: C7306600-15E| Item: 6228825

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to tan liquid
  • Density: 1.61
  • Solubility information: miscible
  • pH range: <0.1
  • 15 gallon drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTextile
  • Concentration70% w/w
  • Linear FormulaH2SO4
  • ClassVolumetric
  • FormLiquid
  • Volume (Gal)15
  • PackagingDrum
  • DOT InformationSULFURIC ACID SOLUTION, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number7664-93-9
  • UN NumberUN2796
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Boiling Point C (° C)165
  • Boiling Point F (° F)329
  • DescriptionSulfuric Acid, 70% (w/w), Textile Grade, Drum, 15 gal.
Documents & Certificates
Reviews
Q&A