Reagents® Sodium Thiosulfate, 0.10N, Standardized

Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/10
  • Assay 0.0995 to 0.1005 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1.01
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 6 to 8.4
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Sodium Thiosulfate, 0.10N, Standardized, Hedpak Container, 20 L 6228840 Mfr # C7328100-20E3
COMPARE
Standardized Hedpak container 20000
Call For Price
Reagents Sodium Thiosulfate, 0.10N, Standardized, Natural Poly Bottle, 4 L 6228841 Mfr # C7328100-4A
COMPARE
Standardized Poly bottle 4000
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat