Reagents Sodium Thiosulfate, 0.10N, Standardized, Hedpak Container, 20 L

Representative image only.

Reagents Sodium Thiosulfate, 0.10N, Standardized, Hedpak Container, 20 L

Reagents | Mfr: C7328100-20E3| Item: 6228840

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • N/10
  • Assay 0.0995 to 0.1005 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1.01
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 6 to 8.4
  • 20 L Hedpack container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Normality0.1
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)20000
  • PackagingHedpak container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionSodium Thiosulfate, 0.10N, Standardized, Hedpak Container, 20 L
Documents & Certificates
Reviews
Q&A