Reagents® Sodium Hydroxide, Lab Grade

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 96%
  • White beads
  • Density: 2.13
  • Solubility information: 1000 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Sodium Hydroxide, Lab Grade, Fiber Drum, 100 lb 6228578 Mfr # C2318250-100F5
COMPARE
Base 45360 Sodium Hydroxide, Lab Grade, Fiber Drum, 100 lb
Call For Price
Reagents Sodium Hydroxide, Lab Grade, Poly Pail, 10 kg 6228579 Mfr # C2318250-10KG
COMPARE
Base 10000 Sodium Hydroxide, Lab Grade, Poly Pail, 10 kg
Call For Price
Reagents Sodium Hydroxide, Lab Grade, Fiber Drum, 25 lb 6228580 Mfr # C2318250-25LB
COMPARE
Base 11340 Sodium Hydroxide, Lab Grade, Fiber Drum, 25 lb
Call For Price
Reagents Sodium Hydroxide, Lab Grade, Natural Poly Container, 3 kg 6228581 Mfr # C2318250-3A5
COMPARE
Base 3000 Sodium Hydroxide, Lab Grade, Natural Poly Container, 3 kg
Call For Price
Reagents Sodium Hydroxide, Lab Grade, Fiber Drum, 50 lb 6228582 Mfr # C2318250-50F5
COMPARE
Base 22680 Sodium Hydroxide, Lab Grade, Fiber Drum, 50 lb
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat