Reagents Sodium Hydroxide, Lab Grade, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Hydroxide, Lab Grade, Poly Pail, 10 kg

Reagents | Mfr: C2318250-10KG| Item: 6228579

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 96%
  • White beads
  • Density: 2.13
  • Solubility information: 1000 g/L at 25 °C (77 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeLab
  • Linear FormulaNaOH
  • ClassBase
  • ChemistryInorganics
  • FormSolid
  • Weight (g)10000
  • Formula Weight40
  • PackagingPoly pail
  • DOT InformationSODIUM HYDROXIDE, SOLID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number1310-73-2
  • UN NumberUN1823
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)1390
  • Boiling Point F (° F)2534
  • Melting Point C (° C)318
  • Melting Point F (° F)604.4
  • DescriptionSodium Hydroxide, Lab Grade, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A