Reagents® Sodium Hydroxide, 0.5N, Standardized

Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/2
  • Assay 0.4995 to 0.5005 Normal
  • Aqueous
  • Colorless liquid
  • Solubility information: miscible
  • pH range: 13.7
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Sodium Hydroxide, 0.5N, Standardized, Cubitainer® Package, 4 L 6228831 Mfr # C7318250-4F
COMPARE
Standardized Cubitainer® package 4000
Call For Price
Reagents Sodium Hydroxide, 0.5N, Standardized, Natural Poly Bottle, 500 mL 6228832 Mfr # C7318250-500A
COMPARE
Standardized Poly bottle 500
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat