Reagents Sodium Hydroxide, 0.5N, Standardized, Natural Poly Bottle, 500 mL

Representative image only.

Reagents Sodium Hydroxide, 0.5N, Standardized, Natural Poly Bottle, 500 mL

Reagents | Mfr: C7318250-500A| Item: 6228832

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • N/2
  • Assay 0.4995 to 0.5005 Normal
  • Aqueous
  • Colorless liquid
  • Solubility information: miscible
  • pH range: 13.7
  • 500 mL natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Normality0.5
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationSODIUM HYDROXIDE SOLUTION, 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1824
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionSodium Hydroxide, 0.5N, Standardized, Natural Poly Bottle, 500 mL
Documents & Certificates
Reviews
Q&A