Reagents® Sodium Dichromate, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Orange-red solid
  • Density: 2.35
  • Boiling point: approximately 400 °C (752 °F), decomposes
  • Melting point: 357 °C (674.6) anhydrous salt
  • Solubility information: 1870 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Sodium Dichromate, Purified, Poly Pail, 10 kg 6228573 Mfr # C2316000-10KG
COMPARE
Salts 10000 Sodium Dichromate, Purified, Poly Pail, 10 kg
Call For Price
Reagents Sodium Dichromate, Purified, Fiber Drum, 50 lb 6228574 Mfr # C2316000-50F5
COMPARE
Salts 22680 Sodium Dichromate, Purified, Fiber Drum, 50 lb
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat