Reagents Sodium Dichromate, Purified, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Dichromate, Purified, Poly Pail, 10 kg

Reagents | Mfr: C2316000-10KG| Item: 6228573

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Orange-red solid
  • Density: 2.35
  • Boiling point: approximately 400 °C (752 °F), decomposes
  • Melting point: 357 °C (674.6) anhydrous salt
  • Solubility information: 1870 g/L at 25 °C (77 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2Cr2O7·2H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsSodium dichromate dihydrate
  • Weight (g)10000
  • Formula Weight298
  • PackagingPoly pail
  • DOT InformationTOXIC SOLID, CORROSIVE, INORGANIC, N.O.S. (SODIUM DICHROMATE), 6.1 (8), P.G. II
  • Storage SymbolAmbient
  • UN NumberUN3290
  • CAS Number7789-12-0
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • DescriptionSodium Dichromate, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A