Reagents® Methyl Orange Indicator, 0.1% (w/v), Volumetric, ASTM D974

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Prepared in accordance with ASTM D974 procedures
  • Orange liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 3.2 (pink) to 4.4 (yellow)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Cubitainer® Package, 10 L 6228680 Mfr # C6118000-10F
COMPARE
Indicator Cubitainer 10000
Call For Price
Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Natural Poly Bottle, 120 mL 6228681 Mfr # C6118000-120A
COMPARE
Indicator Poly bottle 120
Call For Price
Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Cubitainer® Package, 20 L 6228682 Mfr # C6118000-20F
COMPARE
Indicator Cubitainer 20000
Call For Price
Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Natural Poly Bottle, 4 L 6228683 Mfr # C6118000-4A
COMPARE
Indicator Poly bottle 4000
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat