Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Cubitainer® Package, 20 L

Representative image only.

Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Cubitainer® Package, 20 L

Reagents | Mfr: C6118000-20F| Item: 6228682

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Prepared in accordance with ASTM D974 procedures
  • Orange liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 3.2 (pink) to 4.4 (yellow)
  • 20 L Cubitainer package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.1% (w/v)
  • Purity GradeASTM
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number547-58-0, 7732-18-5
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionMethyl Orange Indicator, 0.1% (w/v), Volumetric, Cubitainer® Package, 20 L
Documents & Certificates
Reviews
Q&A