Reagents® Methanol (Absolute), ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99.8%
  • Colorless liquid
  • Slight, alcohol-like odor
  • Density: 0.79
  • Refractive index: 1.328 to 1.33nD
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Methanol, ACS Reagent (Absolute), Amber Glass Bottle, 1 L 6228130 Mfr # C1121000-1C
COMPARE
Solvents Amber glass bottle 1000
Call For Price
Reagents Methanol, ACS Reagent (Absolute), Clear Glass Bottle, 2.5L 6228131 Mfr # C1121000-2.5D1
COMPARE
Solvents Clear glass bottle 2500
Call For Price
Reagents Methanol, ACS Reagent (Absolute), Drum, 200 L 6228132 Mfr # C1121000-200E
COMPARE
Solvents Drum container 200000
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat