Reagents Methanol, ACS Reagent (Absolute), Amber Glass Bottle, 1 L

Representative image only.

Reagents Methanol, ACS Reagent (Absolute), Amber Glass Bottle, 1 L

Reagents | Mfr: C1121000-1C| Item: 6228130

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.8%
  • Colorless liquid
  • Slight, alcohol-like odor
  • Density: 0.79
  • Refractive index: 1.328 to 1.33nD
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCH3OH
  • ClassSolvents
  • ChemistrySolvents
  • SynonymsMethyl Alcohol, Wood Alcohol
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight32.0419
  • Vapor Pressure (torr)233
  • PackagingAmber glass bottle
  • DOT InformationMETHANOL, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-56-1
  • UN NumberUN1230
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)64.5
  • Boiling Point F (° F)148.1
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionMethanol, ACS Reagent (Absolute), Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A