Reagents® Mannitol (D-), ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • White powder
  • Density: 1.52
  • Boiling point: approximately 290 °C (554 °F)
  • Melting point: 167 to 170 °C (332.6 to 338 °F)
  • Solubility information: 180 g/L at 20 °C (68 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Mannitol (D-), ACS Reagent, Natural Poly Container, 1 kg 6228214 Mfr # C1243100-1KG
COMPARE
Salts Poly container 1000
Call For Price
Reagents Mannitol (D-), ACS Reagent, Natural Poly Container, 2 kg 6228215 Mfr # C1243100-2A5
COMPARE
Salts Poly container 2000
Call For Price
Reagents Mannitol (D-), ACS Reagent, Poly Pail, 3 kg 6228216 Mfr # C1243100-3KG
COMPARE
Salts Poly pail 3000
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat