Reagents Mannitol (D-), ACS Reagent, Natural Poly Container, 1 kg

Representative image only.

Reagents Mannitol (D-), ACS Reagent, Natural Poly Container, 1 kg

Reagents | Mfr: C1243100-1KG| Item: 6228214

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White powder
  • Density: 1.52
  • Boiling point: approximately 290 °C (554 °F)
  • Melting point: 167 to 170 °C (332.6 to 338 °F)
  • Solubility information: 180 g/L at 20 °C (68 °F)
  • 1 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaHOCH2(CHOH)4CH2OH
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • SynonymsD-Mannitol
  • Weight (g)1000
  • Formula Weight182.17
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number69-65-8
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionMannitol (D-), ACS Reagent, Natural Poly Container, 1 kg
Documents & Certificates
Reviews
Q&A