Reagents® Ferric Chloride Hexahydrate, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Brownish-orange solid
  • Density: 2.9
  • Boiling point: 280 to 285 °C (536 to 545 °F)
  • Solubility information: 912 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Ferric Chloride Hexahydrate, Purified, Poly Pail, 12 kg 6228504 Mfr # C2204000-12A5
COMPARE
Salts Poly pail 12000
Call For Price
Reagents Ferric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 2 kg 6228505 Mfr # C2204000-2KG
COMPARE
Salts Wide-mouth amber glass bottle 2000
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat