Reagents Ferric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 2 kg

Representative image only.

Reagents Ferric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 2 kg

Reagents | Mfr: C2204000-2KG| Item: 6228505

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Brownish-orange solid
  • Density: 2.9
  • Boiling point: 280 to 285 °C (536 to 545 °F)
  • Solubility information: 912 g/L at 25 °C (77 °F)
  • 2 kg wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaFeCl3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsFerric chloride, Iron (III) chloride, Iron trichloride, Iron(III) chloride hexahydrate
  • Weight (g)2000
  • PackagingWide-mouth amber glass bottle
  • DOT InformationCORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. (FERRIC CHLORIDE, HEXAHYDRATE), 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number10025-77-1
  • UN NumberUN3260
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)37
  • Melting Point F (° F)98.6
  • DescriptionFerric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 2 kg
Documents & Certificates
Reviews
Q&A