Reagents® Deionized Water, ASTM Type IV (D 1193-06)

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Water meets ASTM D 1193 Type IV certification
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Packaging Purity Grade Volume (mL)
Availability Pricing
Reagents Deionized Water, ASTM Type IV (D 1193-06), Cubitainer® Package, 20 L 6228630 Mfr # C2346550-20F
COMPARE
Cubitainer ASTM Type IV 20000
Call For Price
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat