Reagents Deionized Water, ASTM Type IV (D 1193-06), Cubitainer® Package, 20 L

Representative image only.

Reagents Deionized Water, ASTM Type IV (D 1193-06), Cubitainer® Package, 20 L

Reagents | Mfr: C2346550-20F| Item: 6228630

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price

Buy It Together

Current Item

Reagents Deionized Water, ASTM Type IV (D 1193-06), Cubitainer® Package, 20 L
Reagents Deionized Water, ASTM Type IV (D 1193-06), Cubitainer® Package, 20 L
6228630

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Water meets ASTM D 1193 Type IV certification
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • 20 L Cubitainer package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeASTM Type IV
  • Empirical FormulaH2O
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDihydrogen oxide, Dihydrogen monoxide
  • Volume (mL)20000
  • Formula Weight18.02
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • Storage SymbolAmbient
  • CAS Number7732-18-5
  • Shelf Life (days)540
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionDeionized Water, ASTM Type IV (D 1193-06), Cubitainer® Package, 20 L
Documents & Certificates
Reviews
Q&A