Reagents® Citric Acid Monohydrate, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%
  • Colorless/white solid
  • Melting point: 135 to 152 °C (275 to 305.6 °F)
  • Density: 1.54
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Citric Acid Monohydrate, ACS Reagent, Poly Container, 10 kg 6228116 Mfr # C1104500-10KG
COMPARE
Salts Poly container 10000
Call For Price
Reagents Citric Acid Monohydrate, ACS Reagent, Wide-Mouth Natural Poly Bottle, 3 kg 6228117 Mfr # C1104500-3A5
COMPARE
Salts Wide-mouth natural poly bottle 3000
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat