Reagents Citric Acid Monohydrate, ACS Reagent, Wide-Mouth Natural Poly Bottle, 3 kg

Representative image only.

Reagents Citric Acid Monohydrate, ACS Reagent, Wide-Mouth Natural Poly Bottle, 3 kg

Reagents | Mfr: C1104500-3A5| Item: 6228117

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white solid
  • Melting point: 135 to 152 °C (275 to 305.6 °F)
  • Density: 1.54
  • 3 kg wide-mouth natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC6H8O7·H2O
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • SynonymsCitric acid 1-hydrate; 2-hydroxypropane-1,2,3-tricarboxylic acid
  • Weight (g)3000
  • Formula Weight210.14
  • PackagingWide-mouth natural poly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number5949-29-1
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)153
  • Melting Point F (° F)307.4
  • DescriptionCitric Acid Monohydrate, ACS Reagent, Wide-Mouth Natural Poly Bottle; 3kg
Documents & Certificates
Reviews
Q&A