Reagents® Calcium Nitrate Tetrahydrate, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%
  • Colorless/white solid
  • Density: 1.82
  • Solubility information: 2660 g/L at 0 °C (32 °F)
  • pH range: 5 to 7
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Description
Availability Pricing
Reagents Calcium Nitrate Tetrahydrate, ACS Reagent, Fiber Drum, 50 lb 6228160 Mfr # C1165500-50F5
COMPARE
Salts Fiber drum Calcium Nitrate Tetrahydrate, ACS Reagent, Fiber Drum, 50 lb
Call For Price
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat