Reagents Calcium Nitrate Tetrahydrate, ACS Reagent, Fiber Drum, 50 lb

Representative image only.

Reagents Calcium Nitrate Tetrahydrate, ACS Reagent, Fiber Drum, 50 lb

Reagents | Mfr: C1165500-50F5| Item: 6228160

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white solid
  • Density: 1.82
  • Solubility information: 2660 g/L at 0 °C (32 °F)
  • pH range: 5 to 7
  • 50 lb fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCa(NO3)2·4H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)22679.6
  • Formula Weight236.15
  • PackagingFiber drum
  • DOT InformationCALCIUM NITRATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number13477-34-4
  • UN NumberUN1454
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)132
  • Boiling Point F (° F)269.6
  • Melting Point C (° C)561
  • Melting Point F (° F)1041.8
  • DescriptionCalcium Nitrate Tetrahydrate, ACS Reagent, Fiber Drum, 50 lb
Documents & Certificates
Reviews
Q&A