Reagents® Ascorbic Acid

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white solid
  • Density: 1.65
  • Boiling point: 190 °C (374 °F), decomposes
  • Melting point: 190 °C (374 °F), decomposes
  • Solubility information: 330 g/L at 20 °C (68 °F)
  • pH range: 2.4 to 2.8
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Ascorbic Acid, Poly Pail, 12 kg 6228392 Mfr # C2144800-12A5
COMPARE
Salts Poly pail 12000
Call For Price
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat