Reagents Ascorbic Acid, Poly Pail, 12 kg

Representative image only.

Reagents Ascorbic Acid, Poly Pail, 12 kg

Reagents | Mfr: C2144800-12A5| Item: 6228392

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.65
  • Boiling point: 190 °C (374 °F), decomposes
  • Melting point: 190 °C (374 °F), decomposes
  • Solubility information: 330 g/L at 20 °C (68 °F)
  • pH range: 2.4 to 2.8
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaC6H8O6
  • ClassSalts
  • ChemistryOrganics
  • SynonymsL-Ascorbic acid, Vitamin C
  • FormSolid
  • Weight (g)12000
  • Formula Weight176.12
  • Vapor Pressure (torr)7.9179 Pa at 192.15°C
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number50-81-7
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionAscorbic Acid, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A