Reagents® Ammonium Hydroxide, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Assay 28.0 to 30.0%
  • Colorless liquid
  • Density: 0.9
  • Solubility information: miscible
  • pH range: 12.2
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Ammonium Hydroxide, ACS Reagent, Poly Drum, 194 L 6228651 Mfr # C5103500-194Y
COMPARE
Salts Poly drum 194000
Call For Price
Reagents Ammonium Hydroxide, ACS Reagent, Safety Coated Clear Glass Bottle, 2.5 L 6228652 Mfr # C5103500-2.5D
COMPARE
Salts Safety coated clear glass bottle 2500
Call For Price
Reagents Ammonium Hydroxide, ACS Reagent, Clear Glass Bottle, 2.5 L 6228653 Mfr # C5103500-2.5D1
COMPARE
Salts Clear glass bottle 2500
Call For Price
Reagents Ammonium Hydroxide, ACS Reagent, Amber Glass Bottle, 500 mL 6228654 Mfr # C5103500-500C
COMPARE
Salts Amber glass bottle 500
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat