Reagents Ammonium Hydroxide, ACS Reagent, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Ammonium Hydroxide, ACS Reagent, Amber Glass Bottle, 500 mL

Reagents | Mfr: C5103500-500C| Item: 6228654

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Assay 28.0 to 30.0%
  • Colorless liquid
  • Density: 0.9
  • Solubility information: miscible
  • pH range: 12.2
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNH4OH
  • ClassSalts
  • ChemistryInorganics
  • FormLiquid
  • Volume (mL)500
  • Formula Weight35.05
  • PackagingAmber glass bottle
  • DOT InformationAMMONIA SOLUTIONS, 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number1336-21-6
  • UN NumberUN2672
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)24.7
  • Boiling Point F (° F)76.46
  • Melting Point C (° C)-91.5
  • Melting Point F (° F)-132.7
  • DescriptionAmmonium Hydroxide, ACS Reagent, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A