Peter Swan Refrigerators and Freezers for Laboratory Use

Economical cold storage for your laboratory
These general-purpose laboratory refrigerators and freezers are designed to meet most laboratory requirements, with refrigerators maintaining a temperature of 4degC and freezers maintaining a temperature of -20°C.

Research Laboratory refrigerators and freezers are available in either standard duty or heavy duty models. These models are internally spark-free and feature a standard door lock for added protection of samples. The heavy duty models are designed for busy laboratories, incorporating an upgraded refrigeration system with extra thick insulation, assisting in quicker recovery times when door is opened.

Electronic temperature control freezers also feature a standard door lock. Models 44444-60, 44444-62, and 44444-63 feature fixed freezing shelves and are internally spark-free.
Call For Price

38 variations of this product are available.

See All Product Options Below
38 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature - Freezer (° C) Min Temperature - Refrigerator (° C) Refrigerator Capacity (Liters)
Availability Pricing
Peter Swan SW140 RL Refrigerator, 140-liter, 4C, 230V WZ-44444-02 Mfr # SW140 RL
COMPARE
-86
Call For Price
Peter Swan SW180 RL Refrigerator, Heavy-duty 180-liter, 4C, 230V WZ-44444-10 Mfr # SW180 RL
COMPARE
180
Call For Price
Peter Swan SW260 RL Refrigerator, Heavy-duty 260-liter, 4C, 230V WZ-44444-12 Mfr # SW260 RL
COMPARE
260
Call For Price
Peter Swan SW350 RL Refrigerator, Heavy-duty 350-liter, 4C, 230V WZ-44444-14 Mfr # SW350 RL
COMPARE
350
Call For Price
Peter Swan SW250 RLF Refrigerator, 250-liter with recirculating fan, 4C, 230V WZ-44444-22 Mfr # SW250 RLF
COMPARE
Call For Price
Peter Swan SW370 RLF Refrigerator, 370-liter with recirculating fan, 4C, 230V WZ-44444-24 Mfr # SW370 RLF
COMPARE
Call For Price
Peter Swan SW380 RLE Refrigerator, 380-liter digital, 4C, 230V WZ-44444-34 Mfr # SW380 RLE
COMPARE
1 380
Call For Price
Peter Swan SW520 RLE Refrigerator, 520-liter digital, 4C, 230V WZ-44444-35 Mfr # SW520 RLE
COMPARE
2
Call For Price
Peter Swan SW425 SRW Refrigerator, High capacity, 425 liter, 4C, 230V WZ-44444-40 Mfr # SW425 SRW
COMPARE
-6 2 425
Call For Price
Peter Swan SW660 SRS Refrigerator, High capacity 660-liters, 4C, 230V WZ-44444-42 Mfr # SW660 SRS
COMPARE
1 660
Call For Price
Peter Swan SW1270 SRS Refrigerator, High capacity 1270-liters, 4C, 230V WZ-44444-43 Mfr # SW1270 SRS
COMPARE
2
Call For Price
Peter Swan SW1400 SRS Refrigerator, High capacity 1400-liters, 4C, 230V WZ-44444-44 Mfr # SW1400 SRS
COMPARE
2
Call For Price
Peter Swan SW105 VF Freezer, Upright 105-liter, -20C, 230V WZ-44444-51 Mfr # SW105 VF
COMPARE
Call For Price
Peter Swan SW340 VF Freezer, Heavy-Duty Upright 340-liter, -20C, 230V WZ-44444-54 Mfr # SW340 VF
COMPARE
0
Call For Price
Peter Swan SW505 VF Freezer, Heavy-Duty Upright 505-liter, -20C, 230V WZ-44444-55 Mfr # SW505 VF
COMPARE
-20
Call For Price
Peter Swan SW525 VF Freezer, Heavy-Duty Upright 525-liter, -20C, 230V WZ-44444-56 Mfr # SW525 VF
COMPARE
2
Call For Price
Peter Swan SW145 VFE Freezer, Electronic Upright 145-liter, -20C, 230V WZ-44444-61 Mfr # SW145 VFE
COMPARE
Call For Price
Peter Swan SW410 VFE Freezer, Electronic Upright 410-liter, -20C, 230V WZ-44444-63 Mfr # SW410 VFE
COMPARE
-20
Call For Price
Peter Swan SW425 SFW Freezer, High-Capacity Upright 425-liter, -20C, 230V WZ-44444-65 Mfr # SW425 SFW
COMPARE
Call For Price
Peter Swan SW1270 SFS Freezer, High-Capacity Upright 1270-liter, -20C, 230V WZ-44444-68 Mfr # SW1270 SFS
COMPARE
Call For Price
Showing 1 - 20 of 38

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat