Orochem Panthera Deluxe MicroElute 96-Well Plates
Unique well design provides a high sorbent bed and minimum void volume
  • High recovery 96-well collection plates significantly reduce solvent consumption
  • Reproducible packing and recovery
  • Possible for up to 20-fold sample enrichment
The Orochem Panthera Deluxe MicroElute 96-well plates feature a unique well design that offers minimum void volume to achieve maximum solvent recovery and high sorbent bed height to obtain maximum sorbent capacity. These highly sensitive plates are ideal for low sample volume bioanalysis in the pharmaceutical industry, CRO, and clinical diagnostics labs. Plates have a sample loading volume of 10-500 µL and a minimum elution volume of 25 µL.
£188.33 - £210.11GBP / Each
£226.00 - £252.13GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Total Volume Capacity (µL) Methodology
Availability Pricing
Orochem Panthera Deluxe MicroElute SPE 96-Well Plate with Collection Plate, Reversed Phase, 3 mg/Well; 1/EA 6482221 Mfr # OC03PA6002
COMPARE
Reversed Phase 500 Panthera Deluxe
£188.33GBP / Each
£226.00GBP (incl VAT)
Orochem Agility Deluxe MicroElute SPE 96-Well Plate with Collection Plate, WAX, 3 mg/Well; 1/EA 6482222 Mfr # OC03AGD7002
COMPARE
WAX 500 Agility Deluxe
£210.11GBP / Each
£252.13GBP (incl VAT)