Orochem Agility Deluxe MicroElute SPE 96-Well Plate with Collection Plate, WAX, 3 mg/Well; 1/EA
Representative image only.
Orochem Agility Deluxe MicroElute SPE 96-Well Plate with Collection Plate, WAX, 3 mg/Well; 1/EA
Orochem | Mfr: OC03AGD7002| Item: 6482222
Unique well design provides a high sorbent bed and minimum void volume
£210.11GBP / Each
£252.13GBP (incl VAT)
£210.11GBP / Each
£252.13GBP (incl VAT)
Product Description
Key Features
  • High recovery 96-well collection plates significantly reduce solvent consumption
  • Reproducible packing and recovery
  • Possible for up to 20-fold sample enrichment
More About this Item
The Orochem Panthera Deluxe MicroElute 96-well plates feature a unique well design that offers minimum void volume to achieve maximum solvent recovery and high sorbent bed height to obtain maximum sorbent capacity. These highly sensitive plates are ideal for low sample volume bioanalysis in the pharmaceutical industry, CRO, and clinical diagnostics labs. Plates have a sample loading volume of 10-500 µL and a minimum elution volume of 25 µL.
Specifications
  • PhaseIon-exchanged Phase
  • Bottom Typepoly-DVB polymer
  • MethodologyAgility Deluxe
  • SorbentWAX
  • Total Volume Capacity (µL)500
  • Vessel96-Well Plate
  • DescriptionAgility Deluxe MicroElute SPE 96-Well Plate with Collection Plate, WAX, 3 mg/Well; 1/EA