NSI Lab Solutions Quantitative Food Microbiology PT Express™ Cultures
On-demand proficiency test (PT) for when you need results fast
  • Proficiency test for quantitative determination of target microorganisms
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These food microbiology proficiency tests provide the quantitative determination of target microorganisms by standardized cultural methods. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation. If applicable add meat or dairy matrix to the solution. Your PT report is available within 48 hours once you submit your results online.
Food Microbiology PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Call For Price
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description Packaging
Availability Pricing
COMPARE
Quantitative Indicators with Buffer Matrix PT Express™; 2/PK 2/PK
Call For Price
COMPARE
Quantitative Indicators with Buffer Matrix PT Express™; 5/PK 5/PK
Call For Price
COMPARE
Quantitative Indicators with Dairy Matrix PT Express™; 2/PK 2/PK
Call For Price
COMPARE
Quantitative Indicators with Meat Matrix PT Express™; 5/PK 5/PK
Call For Price
COMPARE
Quantitative Lactic Acid Bacteria with Buffer Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative P. aeruginosa with Buffer Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative B. cereus with Dairy Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative Indicators with Meat Matrix PT Express™; 2/PK 2/PK
Call For Price
COMPARE
Quantitative B. cereus with Buffer Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative Indicators with Buffer Matrix PT Express™; 10/PK 10/PK
Call For Price
COMPARE
Quantitative Indicators with Dairy Matrix PT Express™; 10/PK 10/PK
Call For Price
COMPARE
Quantitative Indicators with Meat Matrix PT Express™; 10/PK 10/PK
Call For Price
COMPARE
Quantitative B. cereus with Meat Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative Lactic Acid Bacteria with Meat Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative P. aeruginosa with Meat Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative Lactic Acid Bacteria with Dairy Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative P. aeruginosa with Dairy Matrix PT Express™ 2/PK
Call For Price
COMPARE
Quantitative Indicators with Dairy Matrix PT Express™; 5/PK 5/PK
Call For Price
Showing 1- 18 of 18