NSI Lab Solutions FMPT-001-10B Quantitative Indicators with Buffer Matrix PT Express™; 10/PK

Representative image only.

NSI Lab Solutions FMPT-001-10B Quantitative Indicators with Buffer Matrix PT Express™; 10/PK

NSI Lab Solutions | Item: 6120408

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Includes

10 vials of lyophilized pellet and 10 bottles of buffer.

Key Features

  • Proficiency test for quantitative determination of target microorganisms
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For quantitative determination of aerobic plate count (APC), total coliform, E. coli, S.aureus, Enterobacteriaceae, and yeast/mold in the range of 100 to 25,000 CFU/pellet after hydration
  • Supplied with buffer matrix
  • Pack of 10

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These food microbiology proficiency tests provide the quantitative determination of target microorganisms by standardized cultural methods. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation. If applicable add meat or dairy matrix to the solution. Your PT report is available within 48 hours once you submit your results online.

Food Microbiology PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

When prepared according to the instructions, the concentration range for the following seven analytes will be:

APC: 2,500 to 500,000 CFU/pellet
Total Coliform: 2,000 to 400,000 CFU/pellet
E. coli: 1,000 to 200,000 CFU/pellet
S. aureus: 5,000 to 100,000 CFU/pellet
Enterobacteriaceae: 2,500 to 400,000 CFU/pellet
Yeast: 5,000 to 100,000 CFU/pellet
Mold (if present): 500 to 100,000 CFU/pellet

Specifications
  • OrganismTotal coliforms, E. coli, S. aureus, Enterobacteriaceae, yeast/mold
  • Organism typeBacteria, fungi
  • Packaging10/PK
  • Product TypeFood Microbiology PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQuantitative Indicators with Buffer Matrix PT Express™; 10/PK
Documents & Certificates
Reviews
Q&A