NSI Lab Solutions Pharmaceutical Secondary Reference Materials, A to L

Highly purified compounds—excellent for establishing qualitative and quantitative data
  • Direct traceability of purity and identity to pharmacopeia primary standards established by multiple methods of analysis
  • Great substitute for primary standards for routine quality control purposes
  • ISO 17034 accredited
Pharmaceutical secondary reference materials have been identity and purity certified against multiple analytical methods, including HPLC, FTIR, GC, GC/MS, melting point, and titration. See the Certificate of Analysis for details of the certification process used.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
Call For Price

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Description
Availability Pricing
COMPARE
Ethanol Ethanol (Alcohol) Pharmaceutical Reference Material; 20 mL
Call For Price
COMPARE
Dextrose – Anhydrous Dextrose - Anhydrous Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Caffeine Caffeine Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Ibuprofen Ibuprofen Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Ascorbic Acid Ascorbic Acid Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Fumaric Acid Fumaric Acid Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Calcium Ascorbate Calcium Ascorbate Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Acetaminophen Acetaminophen Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Benzyl alcohol Benzyl alcohol Pharmaceutical Reference Material; 1 g
Call For Price
COMPARE
Diphenhydramine hydrochloride Diphenhydramine Hydrochloride Pharmaceutical Reference Material; 1 g
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat