NSI Lab Solutions PH1476 Dextrose - Anhydrous Pharmaceutical Reference Material; 1 g

Representative image only.

NSI Lab Solutions PH1476 Dextrose - Anhydrous Pharmaceutical Reference Material; 1 g

NSI Lab Solutions | Item: 0500014

Highly purified compounds—excellent for establishing qualitative and quantitative data

Call For Price

Call For Price
Product Description

Key Features

  • Direct traceability of purity and identity to pharmacopeia primary standards established by multiple methods of analysis
  • Great substitute for primary standards for routine quality control purposes
  • ISO 17034 accredited

More About this Item

Pharmaceutical secondary reference materials have been identity and purity certified against multiple analytical methods, including HPLC, FTIR, GC, GC/MS, melting point, and titration. See the Certificate of Analysis for details of the certification process used.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.

Specifications
  • Element/IonDextrose – Anhydrous
  • FormPowder
  • Weight (g)1
  • Storage SymbolStore between 15 and 30 °C
  • CAS Number50-99-7
  • Shelf Life (months)48
  • DescriptionDextrose - Anhydrous Pharmaceutical Reference Material; 1 g
Documents & Certificates
Reviews
Q&A