NSI Lab Solutions Environmental Drinking Water Inorganics Proficiency Tests

Ensure state and accreditation regulation compliance with proficiency testing
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI WS fields of testing
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental chemistry drinking water proficiency tests are designed for identification of target chemicals in drinking water.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
Call For Price

25 variations of this product are available.

See All Product Options Below
25 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
LAS as MBAs 10.5 MBAs Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Color 100 Color Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Silica 21 Silica Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Total Cyanide 21 Cyanide Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Mercury 21 Mercury Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Nitrate 21 Nitrate Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Nitrite 21 Nitrite Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
TOC and DOC 21 TOC/DOC Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Uranium 21 Uranium Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Fluoride 125 Fluoride Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Calcium, Magnesium, Sodium, Calcium Hardness, Total Hardness 250 Hardness Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Vanadium 21 Vanadium Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Turbidity 21 Turbidity Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Alkalinity, Chloride, Conductivity, Fluoride, Nitrate as N, Nitrate/Nitrite to N, Potassium, Total Dissolved Solids 500 Inorganics Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
UV254 absorbance 21 UV254 Asorbance Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Corrosivity 500 Corrosivity Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Perchlorate 5 Perchlorate Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Aluminum, Antimony, Arsenic, Barium, Beryllium, Boron, Cadmium, Chromium, Copper, Iron 21 Trace Metals Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
Fluoride 250 Low Level Fluoride Drinking Water Proficiency Test; 2/PK
Call For Price
COMPARE
pH 250 pH Drinking Water Proficiency Test; 2/PK
Call For Price
Showing 1 - 20 of 25

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat