NSI Lab Solutions Environmental Chlorines Certified Reference Materials
Validate analytical methods and evaluate the quality of your results with certified reference materials
  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Convenient and easy to use—simply dilute 1 mL from an ampule to 1000 mL to reach certified value
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.
£87.00GBP / Pkg of 24
£104.40GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Concentration Description
Availability Pricing
COMPARE
0.050 mg/L Total Residual Chlorine Concentrate CRM, 0.050 mg/L, 2 mL; 24/PK
Call For Price
COMPARE
0.100 mg/L Total Residual Chlorine Concentrate CRM, 0.100 mg/L, 2 mL; 24/PK
Call For Price
COMPARE
1.00 mg/L Total Residual Chlorine Concentrate CRM, 1.00 mg/L, 2 mL; 24/PK
Call For Price
COMPARE
1.50 mg/L Total Residual Chlorine Concentrate CRM, 1.50 mg/L, 2 mL; 24/PK
Call For Price
COMPARE
2.00 mg/L Total Residual Chlorine Concentrate CRM, 2.00 mg/L, 2 mL; 24/PK
£87.00GBP / Pkg of 24
£104.40GBP (incl VAT)
COMPARE
0.500 mg/L Total Residual Chlorine Concentrate CRM, 0.500 mg/L, 2 mL; 24/PK