NSI Lab Solutions QCI-187 Total Residual Chlorine Concentrate CRM, 1.00 mg/L, 2 mL; 24/PK

Representative image only.

NSI Lab Solutions QCI-187 Total Residual Chlorine Concentrate CRM, 1.00 mg/L, 2 mL; 24/PK

NSI Lab Solutions | Item: 0500513

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Convenient and easy to use—simply dilute 1 mL from an ampule to 1000 mL to reach certified value
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • Concentration: 1.00 mg/L

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Specifications
  • Concentration1.00 mg/L
  • Element/IonTotal Residual Chlorine
  • FormLiquid
  • Volume (mL)2
  • Packaging24
  • Storage SymbolRefrigerate between 2 and 8 °C
  • UN Number1791
  • Shelf Life (months)24
  • DescriptionTotal Residual Chlorine Concentrate CRM, 1.00 mg/L, 2 mL; 24/PK
Documents & Certificates
Reviews
Q&A