Labconco Freeze Drying Chambers for FreeZone Freeze Dryers
Expand the flexibility of your freeze drying system with drying chambers
  • Easily attach flasks to your freeze dry system or contain samples in bulk
  • Compatible with all FreeZone Freeze Dryers
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk.
£1,926.86 - £4,034.73GBP / Each
£2,312.23 - £4,841.68GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (InchShort)
Availability Pricing
Labconco FreeZone Freeze Drying Chamber, SS Body and Acrylic Lid, 12 Ports 0334601 Mfr # 7522800
COMPARE
9.75
£2,660.39GBP / Each
£3,192.47GBP (incl VAT)
Labconco FreeZone Freeze Drying Chamber, SS Body with Glass Lid, Short 0334602 Mfr # 7555200
COMPARE
13
£1,926.86GBP / Each
£2,312.23GBP (incl VAT)
Labconco FreeZone Freeze Drying Chamber, SS Body and Acrylic Lid, 16 Ports 0334611 Mfr # 7522900
COMPARE
13
£3,628.17GBP / Each
£4,353.80GBP (incl VAT)
Labconco FreeZone Freeze Drying Chamber, Clear Acrylic Body and Lid, Tall 0335315 Mfr # 7318700
COMPARE
24.625
£3,336.59GBP / Each
£4,003.91GBP (incl VAT)
Labconco FreeZone Freeze Drying Chamber, Clear Acrylic Body and Lid, Short 0335316 Mfr # 7318702
COMPARE
16.875
Call For Price
Labconco FreeZone Freeze Drying Chamber, Clear Acrylic Body and Aluminum Lid, 8 Ports, Short 0335318 Mfr # 7318802
COMPARE
15.25
£4,034.73GBP / Each
£4,841.68GBP (incl VAT)
Labconco FreeZone Freeze Drying Chamber, SS/PTFE-Coated Interior Body and Acrylic Lid, 12 Ports 0335342 Mfr # 7522801
COMPARE
9.75
£3,287.97GBP / Each
£3,945.56GBP (incl VAT)
Showing 1- 7 of 7