Labconco FreeZone Freeze Drying Chamber, SS Body with Glass Lid, Short
Representative image only.
Labconco FreeZone Freeze Drying Chamber, SS Body with Glass Lid, Short
Labconco | Mfr: 7555200| Item: 0334602
Expand the flexibility of your freeze drying system with drying chambers
£1,926.86GBP / Each
£2,312.23GBP (incl VAT)
£1,926.86GBP / Each
£2,312.23GBP (incl VAT)
Product Description
Includes
Clear 3/4" thick glass lid.
Key Features
  • Easily attach flasks to your freeze dry system or contain samples in bulk
  • Compatible with all FreeZone Freeze Dryers
  • Pass-through for electrical cords
  • 8 month warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk.
This stainless steel chamber features a 3/4" glass lid, vacuum release valve, and pass-through for electrical cords. For use with the FreeZone three-shelf sample stand (03353-19).
Specifications
  • Exterior Height (InchShort)13
  • Number of Ports0
  • MaterialSS body with glass lid
  • DescriptionFreeze Drying Chamber, SS Body with Glass Lid, Short
  • Warranty8 Months