HTL Optipette Pipettor Starter Kits
Easily adjust ejector height
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Color Code Description
Availability Pricing
HTL 7902 Optipette Starter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories 2455218 Mfr # 7902
COMPARE
Yes Red, Yellow, and Blue Starter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories
Call For Price
HTL 7912 Optipette Color Starter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories 2455222 Mfr # 7912
COMPARE
Yes Orange, Yellow, Green, and Blue Color Starter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories