HTL 7902 Optipette Starter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories
Representative image only.
HTL 7902 Optipette Starter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories
| Mfr: 7902| Item: 2455218
Easily adjust ejector height
Call For Price
Call For Price
Product Description
Includes
pipettors, stand, and calibration tool.
Key Features
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
  • 3 year warranty
More About this Item
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
3 Year Warranty
Specifications
  • AutoclavableYes
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed, Yellow, and Blue
  • DescriptionStarter Kit with OP10, OP20, OP200 & OP1000 Pipettors and Accessories