HTL Discovery Comfort Pipettor Starter Kits
Comfort, safety, precision and value make this variable volume pipette series quite a discovery
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Convenient kits include everything you need
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and easy-to-calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision.
£546.68 - £745.78GBP / Each
£656.02 - £894.94GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Code Number Of Channels Description
Availability Pricing
HTL 7901 Discovery Comfort Starter Kit with DV10, DV20, DV200 & DV1000 Pipettors and Accessories WZ-24552-17 Mfr # 7901
COMPARE
Red, yellow, and blue 1 Discovery Comfort Starter Kit with DV10, DV20, DV200 & DV1000 Pipettors and Accessories
£745.78GBP / Each
£894.94GBP (incl VAT)
HTL 7904 Discovery Comfort Starter Kit with D10, D20, D200 & D1000 Pipettors and Accessories WZ-24552-20 Mfr # 7904
COMPARE
Red, yellow, and blue 1 Discovery Comfort Starter Kit with D10, D20, D200 & D1000 Pipettors and Accessories
£745.78GBP / Each
£894.94GBP (incl VAT)
HTL 7923 Discovery Comfort Bio Starter Kit with DV10, DV100 & DV1000 Pipettors and Accessories WZ-24552-23 Mfr # 7923
COMPARE
Red, yellow, and blue 1 Discovery Comfort Bio Starter Kit with DV10, DV100 & DV1000 Pipettors and Accessories
£565.06GBP / Each
£678.07GBP (incl VAT)
HTL 7924 Discovery Comfort Starter Kit with DV20, DV200 & DV1000 Pipettors and Accessories WZ-24552-24 Mfr # 7924
COMPARE
Yellow and blue 1 Discovery Comfort Starter Kit with DV20, DV200 & DV1000 Pipettors and Accessories
£546.68GBP / Each
£656.02GBP (incl VAT)
HTL 7925 Discovery Comfort Bio Starter Kit with D10, D100 & D1000 Pipettors and Accessories WZ-24552-25 Mfr # 7925
COMPARE
Red, yellow, and blue 1 Discovery Comfort Bio Starter Kit with D10, D100 & D1000 Pipettors and Accessories
£565.06GBP / Each
£678.07GBP (incl VAT)
HTL 7926 Discovery Comfort Starter Kit with D20, D200 & D1000 Pipettors and Accessories WZ-24552-26 Mfr # 7926
COMPARE
Yellow and blue 1 Discovery Comfort Starter Kit with D20, D200 & D1000 Pipettors and Accessories
£546.68GBP / Each
£656.02GBP (incl VAT)