HTL 7904 Discovery Comfort Starter Kit with D10, D20, D200 & D1000 Pipettors and Accessories
Representative image only.
HTL 7904 Discovery Comfort Starter Kit with D10, D20, D200 & D1000 Pipettors and Accessories
| Mfr: 7904| Item: 2455220
Comfort, safety, precision and value make this variable volume pipette series quite a discovery
£816.75GBP / Each
£980.10GBP (incl VAT)
£816.75GBP / Each
£980.10GBP (incl VAT)
Product Description
Key Features
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Convenient kits include everything you need
  • 3 year warranty
More About this Item
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and easy-to-calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed, yellow, and blue
  • Control Button ColorRed, yellow, and blue
  • DescriptionDiscovery Comfort Starter Kit with D10, D20, D200 & D1000 Pipettors and Accessories