HTL Clinipet+ Fixed-Volume Pipettors
Enjoy accuracy and precision with durable, UV-resistant and autoclavable pipettors
  • Easily removable tip ejector makes it simple to work with narrow tubes
  • Low pipetting forces prevent repetitive strain injury
  • Unique tip sealing system
  • Available in 33 models from 2 µL to 1000 µL
Whether you're right- or left-handed, the Clinipet+ fixed-volume pipettor is smooth to use, thanks to a sliding tip ejector button and smartly designed handle. Durable PVDF shaft guarantees optimum mechanical and chemical resistance. These fixed-volume pipettors also ensure accurate pipette calibration.
Call For Price
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
HTL 2202 Clinipet Series CP2 Fixed-Volume Pipettor, Yellow; 2 µL 2455102 Mfr # 2202
COMPARE
Yellow 2
Call For Price
HTL 2204 Clinipet Series CP5 Fixed-Volume Pipettor, Yellow; 5 µL 2455103 Mfr # 2204
COMPARE
±2 Yellow 5
Call For Price
HTL 2206 Clinipet Series CP7 Fixed-Volume Pipettor, Yellow; 7 µL 2455104 Mfr # 2206
COMPARE
±1.5 Yellow 7
Call For Price
HTL 2208 Clinipet Series CP10 Fixed-Volume Pipettor, Yellow; 10 µL 2455105 Mfr # 2208
COMPARE
±1.2 Yellow 10
Call For Price
HTL 2210 Clinipet Series CP15 Fixed-Volume Pipettor, Yellow; 15 µL 2455106 Mfr # 2210
COMPARE
±1 Yellow 15
Call For Price
HTL 2212 Clinipet Series CP20 Fixed-Volume Pipettor, Yellow; 20 µL 2455107 Mfr # 2212
COMPARE
±0.9 Yellow 20
Call For Price
HTL 2214 Clinipet Series CP25 Fixed-Volume Pipettor, Yellow; 25 µL 2455108 Mfr # 2214
COMPARE
±0.9 Yellow 25
Call For Price
HTL 2216 Clinipet Series CP30 Fixed-Volume Pipettor, Yellow; 30 µL 2455109 Mfr # 2216
COMPARE
±0.9 Yellow 30
Call For Price
HTL 2218 Clinipet Series CP40 Fixed-Volume Pipettor, Yellow; 40 µL 2455110 Mfr # 2218
COMPARE
±0.9 Yellow 40
Call For Price
HTL 2220 Clinipet Series CP44.7 Fixed-Volume Pipettor, Yellow; 44.7 µL 2455111 Mfr # 2220
COMPARE
±0.9 Yellow 44.7
Call For Price
HTL 2222 Clinipet Series CP50 Fixed-Volume Pipettor, Yellow; 50 µL 2455112 Mfr # 2222
COMPARE
±0.9 Yellow 50
Call For Price
HTL 2224 Clinipet Series CP60 Fixed-Volume Pipettor, Yellow; 60 µL 2455113 Mfr # 2224
COMPARE
±0.9 Yellow 60
Call For Price
HTL 2226 Clinipet Series CP70 Fixed-Volume Pipettor, Yellow; 70 µL 2455114 Mfr # 2226
COMPARE
±0.9 Yellow 70
Call For Price
HTL 2228 Clinipet Series CP75 Fixed-Volume Pipettor, Yellow; 75 µL 2455115 Mfr # 2228
COMPARE
±0.9 Yellow 75
Call For Price
HTL 2230 Clinipet Series CP80 Fixed-Volume Pipettor, Yellow; 80 µL 2455116 Mfr # 2230
COMPARE
±0.9 Yellow 80
Call For Price
HTL 2232 Clinipet Series CP90 Fixed-Volume Pipettor, Yellow; 90 µL 2455117 Mfr # 2232
COMPARE
±0.8 Yellow 90
Call For Price
HTL 2234 Clinipet Series CP100 Fixed-Volume Pipettor, Yellow; 100 µL 2455118 Mfr # 2234
COMPARE
±0.8 Yellow 100
Call For Price
HTL 2236 Clinipet Series CP120 Fixed-Volume Pipettor, Yellow; 120 µL 2455119 Mfr # 2236
COMPARE
±0.8 Yellow 120
Call For Price
HTL 2238 Clinipet Series CP150 Fixed-Volume Pipettor, Yellow; 150 µL 2455120 Mfr # 2238
COMPARE
±0.7 Yellow 150
Call For Price
HTL 2240 Clinipet Series CP200 Fixed-Volume Pipettor, Yellow; 200 µL 2455121 Mfr # 2240
COMPARE
±0.6 Yellow 200
Call For Price
Showing 1 - 20 of 33