HTL 2230 Clinipet Series CP80 Fixed-Volume Pipettor, Yellow; 80 µL
Representative image only.
HTL 2230 Clinipet Series CP80 Fixed-Volume Pipettor, Yellow; 80 µL
| Mfr: 2230| Item: 2455116
Enjoy accuracy and precision with durable, UV-resistant and autoclavable pipettors
Call For Price
Call For Price
Product Description
Key Features
  • Easily removable tip ejector makes it simple to work with narrow tubes
  • Low pipetting forces prevent repetitive strain injury
  • Unique tip sealing system
  • Available in 33 models from 2 µL to 1000 µL
  • 3 year warranty
More About this Item
Whether you're right- or left-handed, the Clinipet+ fixed-volume pipettor is smooth to use, thanks to a sliding tip ejector button and smartly designed handle. Durable PVDF shaft guarantees optimum mechanical and chemical resistance. These fixed-volume pipettors also ensure accurate pipette calibration.
3 Year Warranty
Specifications
  • Max Volume (µL)80
  • AutoclavableYes
  • Accuracy (%)±0.9
  • Precision (%)±0.3
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeYellow
  • DescriptionFixed-Volume Pipettor, Yellow; 80 µL