Hamilton® Gastight® Cemented Needle Syringes with Blunt Tip
Blunt stainless steel (SS) needle point—ideal for HPLC injection valves and for sample pipetting
  • Remain gas- and liquid-tight!
  • Easily handles corrosive gases and liquids, radioactive materials, and sterile solutions
  • Epoxy-cemented SS needle provides durability
  • Dead volume is limited to the internal volume of the needle
Hamilton® Gastight® syringes with cemented needle are constructed with a chemically inert glass barrel. Syringes feature a precision-machined PTFE plunger tip to create a leak-free seal, making them ideal for both liquids and gases.
Syringes include an epoxy-cemented SS needle. The "s" on the needle gauge represents a smaller inner diameter for the needle and a thicker needle wall for better durability.
£42.97 - £81.06GBP / Each
£51.56 - £97.27GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Syringe Capacity (µL)
Availability Pricing
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 10 µL, 26s G 0794009 Mfr # 80075
COMPARE
26s 10
£60.80GBP / Each
£72.96GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 10 µL, 26 G 0794011 Mfr # 80085
COMPARE
26 10
£42.97GBP / Each
£51.56GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 25 µL, 22s G 0794017 Mfr # 80275
COMPARE
22s 25
£81.06GBP / Each
£97.27GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 25 µL, 22 G 0794019 Mfr # 80285
COMPARE
22 25
£47.26GBP / Each
£56.71GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 50 µL, 22s G 0794027 Mfr # 80975
COMPARE
22s 50
£49.86GBP / Each
£59.83GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 50 µL, 22 G 0794029 Mfr # 80985
COMPARE
22 50
£47.26GBP / Each
£56.71GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 100 µL, 22s G 0794037 Mfr # 81075
COMPARE
22s 100
£76.46GBP / Each
£91.75GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 100 µL, 22 G 0794039 Mfr # 81085
COMPARE
22 100
£47.26GBP / Each
£56.71GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 250 µL, 22s G 0794047 Mfr # 81175
COMPARE
22s 250
£78.30GBP / Each
£93.96GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 250 µL, 22 G 0794049 Mfr # 81185
COMPARE
22 250
£49.86GBP / Each
£59.83GBP (incl VAT)
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 500 µL, 22 G 0794051 Mfr # 81216
COMPARE
22 500
£76.46GBP / Each
£91.75GBP (incl VAT)
Showing 1- 11 of 11