Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 500 µL, 22 G
Representative image only.
Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 500 µL, 22 G
Hamilton | Mfr: 81216| Item: 0794051
Blunt stainless steel (SS) needle point—ideal for HPLC injection valves and for sample pipetting
£76.46GBP / Each
£91.75GBP (incl VAT)
£76.46GBP / Each
£91.75GBP (incl VAT)
Product Description
Key Features
  • Remain gas- and liquid-tight!
  • Easily handles corrosive gases and liquids, radioactive materials, and sterile solutions
  • Epoxy-cemented SS needle provides durability
  • Dead volume is limited to the internal volume of the needle
  • 500 µL capacity
  • 22 gauge needle
  • 1 year warranty
More About this Item
Hamilton® Gastight® syringes with cemented needle are constructed with a chemically inert glass barrel. Syringes feature a precision-machined PTFE plunger tip to create a leak-free seal, making them ideal for both liquids and gases.
Syringes include an epoxy-cemented SS needle. The "s" on the needle gauge represents a smaller inner diameter for the needle and a thicker needle wall for better durability.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Syringe TypeLuer-Tip Cemented Needle
  • Syringe Capacity (µL)500
  • Needle IncludedYes
  • Needle Tip Style90° Blunt
  • Needle Tip Number3
  • Needle Gauge22
  • Needle Length (in)2
  • MaterialSST, Borosilicate glass, PTFE
  • SterileNo
  • Outside Diameter (OD) (mm)7.75
  • DescriptionSyringe with Cemented Needle, Blunt Tip, 500 µL, 22 G