Extech Controllers
Fuzzy Logic PID, Auto Tuning, and Soft Start features combine for uncompromising, state-of-the-art control
  • Dual 4-digit LED displays for process and setpoint values
  • 1/16 DIN and 1/4 DIN model dimensions available
  • Easy programming & navigation with user-friendly menus and tactile keypad
  • Fuzzy Logic PID offers intuitive control simulating human control logic
  • Manual mode allows the user to override automatic control
  • One-touch Auto Tuning for quick setup and stable, precise control
  • Two ‘Latching’ Alarm relays standard with 8 Alarm modes plus advanced Timer modes
  • Single stage Ramp and Soak program with Ramp-to-Setpoint Limit
  • Accepts thermocouple and RTD inputs
  • Select desired temperature display units (°F or °C) from setup menu
  • Select thermocouple input type (9 selections) or RTD input (2 selections) from the display menu Complete with mounting bracket hardware and screw terminals for easy wiring
Extech controllers offer Fuzzy Logic enhanced PID and a Soft-Start feature that protects heaters from cold-starts. PID plus Fuzzy Logic tackles even the most demanding applications, eliminating over-shoot, unwanted process fluctuations, and drift. The Soft Start feature is ideally suited for processes, such as those employed in the Thermo-Plastics industry, where careful, exact, and slow heating of product is required.
£202.43 - £322.73GBP / Each
£242.92 - £387.28GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
DIN Size Max Temperature (° F) Power (VAC)
Availability Pricing
Extech 48VFL11 1/16 DIN PID Temperature controller with one relay output 8980203 Mfr # 48VFL11
COMPARE
1/16 122 90 to 264
£202.43GBP / Each
£242.92GBP (incl VAT)
Extech 48VFL13 1/16 DIN Temperature PID Controller with 4-20mA Output 8980204 Mfr # 48VFL13
COMPARE
1/16 122 90 to 264
Call For Price
Extech 96VFL11 1/4 DIN Temperature PID Controller with Two Relay Outputs 8980205 Mfr # 96VFL11
COMPARE
1/4 122 90 to 264
£322.73GBP / Each
£387.28GBP (incl VAT)
Extech 96VFL13 1/4 DIN Temperature PID Controller with 4-20mA Output 8980206 Mfr # 96VFL13
COMPARE
1/4 122 90 to 264